1-Benzyl-3-pyrroline 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1-Benzyl-2,5-dihydropyrrole

L19496.03 L19496.06
L19496.03EA 420 DKK
L19496.03 L19496.06
1-Benzyl-3-pyrroline 98%
1-Benzyl-3-pyrroline

Purity: 98+% CAS No.: 6913-92-4 Molecular Formula: C11H13N Formula Weight: 159.23 Storage Temperature: 2°C to 8°C Flash Point: 92°(197°F) Refractive Index: 1.5420 Boiling Point: 60°/1mm Beilstein Reference: 111101 UN No.: UN2735 MDL No.: MFCD00012324

Formel: C₁₁H₁₃N
Kogepunkt: 60 °C (1 mmHg)
Massefylde: 0,837
Flammepunkt: 92 °C (197 °F)
MDL Number: MFCD00012324
CAS-nummer: 6913-92-4
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR