(S)-(-)-3-Acetamidopyrrolidine ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

L12927.03 L12927.06
L12927.03EA 753 DKK
L12927.03 L12927.06
(S)-(-)-3-Acetamidopyrrolidine ≥98%
(S)-(-)-3-Acetamidopyrrolidine

Purity: 98% CAS No.: 114636-31-6 Molecular Formula: C6H12N2O Formula Weight: 128.17 Melting Point: 52-57° Boiling Point: 70-72°/40mm UN No.: 3259 MDL No.: MFCD00082748

Formel: C₆H₁₂N₂O
Molekylvægt: 128.17 g/mol
Kogepunkt: 70…72 °C (40 mmHg)
Smeltepunkt: 52…57 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00082748
CAS-nummer: 114636-31-6
ADR: 8 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR