Sodium hydrogen-L-tartrate, anhydrous 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Sodium hydrogen tartrate, Sodium bitartrate, L-Tartaric acid sodium salt

L12469.22 L12469.36 L12469.14
L12469.22EA 354 DKK
L12469.22 L12469.36 L12469.14
Sodium hydrogen-L-tartrate, anhydrous 98%
Sodium hydrogen-L-tartrate

Purity: 98% CAS No.: 526-94-3 Molecular Formula: C4H5NaO6 Formula Weight: 172.07 Melting Point: ca 243° dec. Beilstein Reference: 6099125 Optical Rotation: +24.5° (c=3 in water) MDL No.: MFCD00065393

Formel: C₄H₅NaO₆
Molekylvægt: 172.07 g/mol
Smeltepunkt: ∼ 243 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00065393
CAS-nummer: 526-94-3
EINECS: 208-400-9
Merck Index: 14,8589

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR