Cyclopropyl phenyl sulphide ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Cyclopropyl(phenyl)sulphane

L12321.03 L12321.06
L12321.03EA 340 DKK
L12321.03 L12321.06
Cyclopropyl phenyl sulphide ≥98%
Cyclopropyl phenyl sulphide

Purity: 98+% CAS No.: 14633-54-6 Molecular Formula: C9H10S Formula Weight: 150.24 Flash Point: 91°(195°F) Refractive Index: 1.5810 Odour: Stench Boiling Point: 61-62°/1mm Beilstein Reference: 2075701 UN No.: 3334 MDL No.: MFCD00009684

Formel: C₉H₁₀S
Molekylvægt: 150.24 g/mol
Kogepunkt: 61…62 °C (1 mmHg)
Massefylde: 1.045
Flammepunkt: 91 °C (195 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009684
CAS-nummer: 14633-54-6
ADR: 9 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR