3-(4-Chlorobenzoyl)acrylic acid 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (2E)-4-(4-Chlorophenyl)-4-oxobut-2-enoic acid

L11467.06 L11467.14
L11467.06EA 363 DKK
L11467.06 L11467.14
3-(4-Chlorobenzoyl)acrylic acid 99%
3-(4-Chlorobenzoyl)acrylic acid

Purity: 99% CAS No.: 6269-33-6 Molecular Formula: C10H7ClO3 Formula Weight: 210.62 Melting Point: 157-159° Beilstein Reference: 2719881 MDL No.: MFCD00041465

Formel: C₁₀H₇ClO₃
Smeltepunkt: 157…159 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041465
CAS-nummer: 6269-33-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR