Cell detachment solution, Accutase
Leverandør: Biowest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L0950-100
L0950-100EA 760 DKK
L0950-100
Cell detachment solution, Accutase
Celledyrkningssupplementer og additiver
Accustase is a ready to use cell detachment solution of proteolytic and collagenolytic enzymes. Accutase is a direct gentle replacement for trypsin solution. It can also be used on suspension cells to reduce their agglutination in order to ease counting.