4,4'-Dibrombenzophenon ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Bis(4-bromophenyl) ketone

L08200.06 L08200.03
L08200.06EA 2740 DKK
L08200.06 L08200.03
4,4'-Dibrombenzophenon ≥98%
4,4'-Dibrombenzophenon

Purity: 98+% CAS No.: 3988-03-2 Molecular Formula: C13H8Br2O Formula Weight: 340.03 Melting Point: 172-176° Beilstein Reference: 2049958

Formel: C₁₃H₈Br₂O
Molekylvægt: 340.01 g/mol
Smeltepunkt: 172…176 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00016330
CAS-nummer: 3988-03-2
EINECS: 223-632-0

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR