5-Methylisatin ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5-Methylindoline-2,3-dione, 5-Methyl-1H-indole-2,3-dione

L08009.03 L08009.06 L08009.14
L08009.03EA 445 DKK
L08009.03 L08009.06 L08009.14
5-Methylisatin ≥97%
5-Methylisatin

Purity: 97% CAS No.: 608-05-9 Molecular Formula: C9H7NO2 Formula Weight: 161.16 Melting Point: ca 180° dec. Beilstein Reference: 123738

Formel: C₉H₇NO₂
Smeltepunkt: ∼ 180 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005721
CAS-nummer: 608-05-9
EINECS: 210-152-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR