7-Azaindol ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1H-Pyrrolo[2,3-b]pyridine

L07983.03 L07983.06 L07983.14
L07983.03EA 476 DKK
L07983.03 L07983.06 L07983.14
7-Azaindol ≥98%
7-Azaindol

Purity: 98% CAS No.: 271-63-6 Molecular Formula: C7H6N2 Formula Weight: 118.14 Melting Point: 103-107° Boiling Point: 270° MDL No.: MFCD00005606

Formel: C₇H₆N₂
Kogepunkt: 270 °C
Smeltepunkt: 103…107 °C
Massefylde: 1,301 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005606
CAS-nummer: 271-63-6
EINECS: 205-981-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR