Perylen ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

L03047.03 L03047.06
L03047.03EA 469 DKK
L03047.03 L03047.06
Perylen ≥98%
Perylene

Purity: 98+% CAS No.: 198-55-0 Molecular Formula: C20H12 Formula Weight: 252.32 Odour: Odourless Melting Point: 276-280° Beilstein Reference: 1911335 Merck Reference: 14,7184 MDL No.: MFCD00004142

Formel: C₂₀H₁₂
Kogepunkt: 443 °C (1013 hPa)
Smeltepunkt: 276…280 °C
Massefylde: 1,362 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004142
CAS-nummer: 198-55-0
EINECS: 205-900-9
Merck Index: 14,7184

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR