Syringe for vapour pressure osmometer
Leverandør: KNAUER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A0433
KNAUA0433EA 5970 DKK
KNAUA0433
Syringe for vapour pressure osmometer
Sprøjter Sprøjter til kromatografi
This syringe has a glass barrel, stainless steel piston with PTFE seal, including needle with spring stopper.