Tubing, PTFE
Leverandør: KNAUER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A0732 A0154 A0873
KNAUA0732EA 445 DKK
KNAUA0732 KNAUA0154 KNAUA0873
Tubing, PTFE
Slanger Overførselsslange
This white PTFE tubing is characterised by its non stick surface and excellent dielectric properties.
  • Excellent chemical resistance
  • Excellent thermal resistance, autoclavable
  • Free of extractable substances, physiologically neutral, biocompatible