Volumetric flasks, with divided neck
Leverandør: Kavalier
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
632431560830
KAVA632431560830EA 432 DKK
KAVA632431560830
Volumetric flasks, with divided neck
Kolber Volumetriske kolber
Borosilicate glass, clear.
  • Blue markings
  • Autoclavable at 121 °C