Heavy-Duty SMS/Film Steam Sterilisation Tubings
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TUBRPKC300-24 TUBRPKC300-12 TUBRPKC300-4 TUBRPKC300-6 TUBRPKC300-9 TUBRPKC300-14 TUBRPKC300-18 TUBRPKC300-28
KACCTUBRPKC300-24EA 19250 DKK
KACCTUBRPKC300-24 KACCTUBRPKC300-12 KACCTUBRPKC300-4 KACCTUBRPKC300-6 KACCTUBRPKC300-9 KACCTUBRPKC300-14 KACCTUBRPKC300-18 KACCTUBRPKC300-28
Heavy-Duty SMS/Film Steam Sterilisation Tubings
Poser Steriliseringsposer
Ideal for applications where the use of standard SMS wraps prevents the user from being able to see and identify the contents of the package.
  • 50 g SMS weight
  • The tubing form allows for sizing flexibility
  • The ends of the tubing should be heat sealed closed
  • When opened, virtually no particulates are generated
  • Manufactured in a certified ISO6 cleanroom
Made from spunbond/meltblown/spunbond polypropylene fabric and a polyester-based flexible film. The SMS side is an effective bacterial barrier, post sterilisation. The clear film side offers excellentMore Product Information