Super-Duty 88 GSM SMS Autoclavable Bowl Covers with Steam Indicator
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BWLKCRP60020 BWLKCRP60024 BWLKCRP60030 BWLKCRP60040 BWLKCRP60060 BWLKCRP60048 BWLKCRP60050 BWLKCRP60036 BWLKCRP60054 BWLKCRP60042
129-4018EA 13810 DKK
129-4018 129-4009 129-4015 129-4017 129-4010 KACCBWLKCRP60048 KACCBWLKCRP60050 KACCBWLKCRP60036 KACCBWLKCRP60054 KACCBWLKCRP60042
Super-Duty 88 GSM SMS Autoclavable Bowl Covers with Steam Indicator
Covers
This 88 g/m² SMS autoclavable bowl cover provides a microbial barrier that is ideal for pharmaceutical production applications.
  • Hydrophobic, tear-resistant, and puncture-resistant
  • Creates secondary level of protection during aseptic presentation and setup of equipment
  • Designed for autoclave cycles (121 to 135 °C)
  • Manufactured and packed in a certified ISO 8 cleanroom
  • Made in USA
Eliminate the need for cutting sheets of sterilisation wrapping materials and securing them with autoclave tape and rubber bands. After the outer bag is removed, thisMore Product Information