Hydroxyethyl cellulose, high purity

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Hydroxyethyl Cellulose

K391-500GEA 4810 DKK
K391-500G
Hydroxyethyl cellulose, high purity
2-Hydroxyethylcellulose
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00072770
CAS-nummer: 9004-62-0

Bestil nu

Specifikationer Testresultater

pH (2%, water) @25 °C 6 – 7


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR