VWR®, DNA ladder, 100 bp
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
K180-250ULEA 1630 DKK
K180-250UL
VWR®, DNA ladder, 100 bp
Elektroforese reagenser Molekylærvægt markører
Ladder with 13 fragments ranging from 100 to 3000 bp for easy band identification.
Also available pre-mixed with loading dye.