9100 Safety goggle, Stealth™
Leverandør: JSP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AGS020-54N-800
JSPAAGS020-54N-800EA 239 DKK
JSPAAGS020-54N-800
9100 Safety goggle, Stealth™
Beskyttelsesbriller
A soft dual injection goggle providing a comfortable universal fit with a high level of protection from splash, dust, and impact.
  • Anti-mist and anti-scratch lens
  • TPV vented inner frame
  • Grade 1 optics lens
  • Ultra-soft housing
  • Wide elastic headband with easy quick release clipping buckle
Large, injection moulded spherical wraparound optically perfect lens provides a clear field of vision. With anti-mist, anti-scratch, and Grade 1 optics. Clear lens also blocks 99.9% of ultraviolet radiation. Ultra-soft housing and dual injection frame provides a softMore Product Information