Vand, Ultrapure for molecular biology nuclease-free

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Hydrogen oxide

J71786.K2 J71786.XX J71786.AE J71786.K8 J71786.AP
J71786.K2EA 725 DKK
J71786.K2 J71786.XX J71786.AE J71786.K8 J71786.AP
Vand, Ultrapure for molecular biology nuclease-free
Vand

Nuclease free water is 0,2 μm filtered and dispensed into vials and/or durable, square Nalgene bottles.


Emballering: The 5 litre size is supplied in an upright carton with a convenient dispensing spout.

Formel: H₂O
Molekylvægt: 18.02 g/mol
Kogepunkt: 100 °C
Smeltepunkt: 0 °C
Massefylde: 1.000
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011332
CAS-nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 14,10039

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR