Netilmicin sulphate 595 µg/mg

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (2R,3R,4R,5R)-2-(((1S,2S,3R,4S,6R)-4-Amino-3-(((2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl)oxy)-6-(ethylamino)-2-hydroxycyclohexyl)oxy)-5-methyl-4-(methylamino)tetrahydro-2H-pyran-3,5-diol sulphate(2:5)

J66302.MA J66302.MB
J66302.MAEA 1850 DKK
J66302.MA J66302.MB
Netilmicin sulphate 595 µg/mg
Netilmicin sulphate

Netilmicin sulphate is used as a broad spectrum antibiotic active against most gram-negative and some gram-positive bacteria.


  • Light Sensitive
  • Hygroscopic


It is a semisynthetic 1-N-ethyl derivative of sisomycin, an aminoglycoside antibiotic with action similar to gentamicin, but has less ear and kidney toxicity. It is used in aminoglycoside resistance studies.


White to off-white powder


Soluble in water


Advarsel: For research use only. Not intended for animal or human diagnostic or therapeutic use.

Formel: C₂₁H₄₁N₅O₇·⁵/₂H₂SO₄
Molekylvægt: 720.75 g/mol
Kogepunkt: 868 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01311715
CAS-nummer: 56391-57-2
EINECS: 260-147-3
Merck Index: 14,6479

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR