Buffer, tris(hydroxymethyl)aminomethane hydrochloride, 1 M stock soln, pH 8,0

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

J63636.K2
J63636.K2EA 1050 DKK
J63636.K2
Buffer, tris(hydroxymethyl)aminomethane hydrochloride, 1 M stock soln, pH 8,0
Buffere Kloning buffere

CAS No.: 1185-53-1
Physical form: liquid
Appearance: colourless
MDL No.: MFCD00012590

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR