Buffer kit for antibody purification on protein A column

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

J63014.QAEA 848 DKK
J63014.QA
Buffer kit for antibody purification on protein A column
Buffere Antistof oprensningsbuffere

Physical Form: Liquid


Applications: Buffers to be used when performing antibody purification using immobilized Protein A.


Leveringsinformation: This kit contains: 1 litre of 1 M Tris-HCl (pH 8.0) and 500 ml of 100 mM glycine (pH 3.0).

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR