EDTA cell culture reagent

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II

J62948.36 J62948.A1
J62948.36EA 1080 DKK
J62948.36 J62948.A1
EDTA cell culture reagent
EDTA

CAS No.: 60-00-4 Molecular Formula: C10H16N2O8 Formula Weight: 292.24 Physical Form: Powder Appearance: White Melting Point: ca 245° dec. Beilstein Reference: 1716295 Merck Reference: 14,3517 MDL No.: MFCD00003541

Formel: C₁₀H₁₆N₂O₈
Molekylvægt: 292.25 g/mol
Kogepunkt: 614
Smeltepunkt: ∼ 245 °C (dec.)
Massefylde: 0.86 g/cm³ (20 °C)
Flammepunkt: >100 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003541
CAS-nummer: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 14,3517
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR