Magnesiumchlorid hexahydrat cell culture reagent

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Magnesium dichloride

J62575.30 J62575.36 J62575.A1
J62575.30EA 756 DKK
J62575.30 J62575.36 J62575.A1
Magnesiumchlorid hexahydrat cell culture reagent
Magnesiumchlorid hexahydrat

CAS No.: 7791-18-6 Molecular Formula: MgCl2.6H2O Formula Weight: 203.30 (95.22anhy) Physical Form: Powder Appearance: White Odour: Odourless Merck Reference: 14,5662 MDL No.: MFCD00149781

Formel: MgCl₂·6H₂O
Molekylvægt: 203.3 g/mol
Kogepunkt: 1412
Smeltepunkt: 117
Massefylde: 1.57 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149781
CAS-nummer: 7791-18-6
EINECS: 232-094-6
Merck Index: 14,5662

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR