Nicotinamid-adenin dinucleotid (oxideret form) 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Nadide, NAD+, Nicotinamide adenine dinucleotide, oxidized

J62337.03 J62337.06 J62337.14
J62337.03EA 662 DKK
J62337.03 J62337.06 J62337.14
Nicotinamid-adenin dinucleotid (oxideret form) 97%
Nicotinamid-adenin dinucleotid (oxideret form)

Purity: 98+% CAS No.: 53-84-9 Molecular Formula: C21H27N7O14P2 Formula Weight: 663.43 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Solubility: Soluble in water at 50mg/ml Beilstein Reference: 3584133 MDL No.: MFCD00036253

Formel: C₂₁H₂₇N₇O₁₄P₂
Molekylvægt: 663.43 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00036253
CAS-nummer: 53-84-9
EINECS: 200-184-4
Merck Index: 14,6344

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR