BIS-TRIS, 0,2 M Buffer Solutions

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

J60127.AE J60127.AK J62209.AE J62209.AK J62686.AE J62686.AK J63346.AE J63346.AK
J60127.AEEA 982 DKK
J60127.AE J60127.AK J62209.AE J62209.AK J62686.AE J62686.AK J63346.AE J63346.AK
BIS-TRIS, 0,2 M Buffer Solutions
Buffere Biologiske buffere

CAS No.: 6976-37-0
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00002853

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR