Natriumpyruvat cell culture reagent

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

J61840.18 J61840.22
J61840.18EA 1530 DKK
J61840.18 J61840.22
Natriumpyruvat cell culture reagent
Natriumpyruvat

CAS No.: 113-24-6 Molecular Formula: C3H3NaO3 Formula Weight: 110.04 Physical Form: Powder Appearance: White to pale yellow Solubility: Freely soluble in water Beilstein Reference: 3568341 MDL No.: MFCD00002586

Formel: CH₃COCOONa
Smeltepunkt: 220…230 °C
Massefylde: 1,718 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002586
CAS-nummer: 113-24-6
EINECS: 204-024-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR