Formaldehyd 4% in PBS stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

Stabilization: Stabilised with methanol

J60401.AK J60401.AP
J60401.AKEA 839 DKK
J60401.AK J60401.AP
Formaldehyd 4% in PBS stabilised
Formaldehyd

Storage Temperature: 2°C to 8°C Appearance: Colourless

Formel: CH₂O
Molekylvægt: 30.03 g/mol
Kogepunkt: -19.3
Smeltepunkt: -92
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00003274
CAS-nummer: 50-00-0
Merck Index: 13,04259

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR