Diethyl pyrocarbonat ≥97%, Ultrapure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC

J14710.AEEA 5360 DKK
J14710.AE J14710.AC
Diethyl pyrocarbonat ≥97%, Ultrapure
Diethyl pyrocarbonat

A strong, non-specific inhibitor of ribonuclease. Reacts with the N1 of histidine residues in proteins under mild conditions. It can be used to cross-link and modify proteins by ethoxyformylation of NH₂ and imidazole groups in proteins. It reacts with amines and thus cannot be used with Tris buffer.

Formel: O(COOC₂H₅)₂
Molekylvægt: 162.14 g/mol
Kogepunkt: 93…94 °C (18 mmHg)
Smeltepunkt: 69 °C
Massefylde: 1.101
Flammepunkt: 69 °C (156 °F)
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00009106
CAS-nummer: 1609-47-8
EINECS: 216-542-8
Merck Index: 14,07998

Bestil nu

Specifikationer Testresultater

Assay (unspecified) ≥97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR