tempmate® S1 Single Use Temperature Data Logger, L1 2 °C, H1 8 °C, SD: 15 min
Leverandør: tempmate
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S1C10A39010
IMECS1C10A39010EA 3620 DKK
IMECS1C10A39010
tempmate® S1 Single Use Temperature Data Logger, L1 2 °C, H1 8 °C, SD: 15 min
Dataloggere
Pre-programmed. Outstanding single-use temperature logger for reliable cold chain monitoring. Integrated USB port and PDF andCSV generating software on-board.
  • Pre-programmed
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food grade packing
  • No additional hardware or software required
  • Detailed PDF and CSV report including graph and table
  • Intelligent LEDs for status monitoring
  • Extended shelf-life of two years
tempmate.®-S1 loggers are specially designed for temperature monitoring in any cold chain logistics. As a pre-More Product Information