Magic LAB® Homogeniser with DR 2000/03 Module
Leverandør: IKA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MAGICLAB
IKAIMAGICLABEA 0 DKK
IKAIMAGICLAB
Magic LAB® Homogeniser with DR 2000/03 Module
Homogenisatorer Homogenisatorer, bordmodeller
The magic LAB® is a unique and multi-functional small-scale laboratory machine. It is designed for mixing, dispersing, wet milling and incorporation of powders into liquids. The magic LAB® is most frequently used for the development of new products or for optimizing of existing process techniques, particularly in the chemical, cosmetic, pharmaceutical and food industries.
  • Depending on the application, the basic unit can be modified quickly with different working modules
  • Flexibility and ease of use: one machine suits for many applications and variety of