Homogeniser, T 25 easy clean control ULTRA-TURRAX®
Leverandør: IKA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25002500
IKAA25002500EA 25260 DKK
IKAA25002500
Homogeniser, T 25 easy clean control ULTRA-TURRAX®
Homogenisatorer Homogenisatorer, bordmodeller
The T 25 easy clean control is a high-performance dispersing instrument with electronic speed control from 3000 to 25000 rpm for working volumes of 1 to 2000 ml (H2O). The S 25 EC-T-C-18G-ST tool allows temperature measurement and operation time detection through RFID. Device overheating is prevented by continuous temperature monitoring and the setting of temperature limits. The detection of operating time serves to guarantee regular maintenance.
  • Continuous temperature measurement with S 25 EC-T-C-18G-ST tool including an integrated temperature sensor