Rotational viscometers, ROTAVISC HELI complete series
Leverandør: IKA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10013698 10013745 10013764 10013731
531-1475EA 32060 DKK
531-1475 IKAA10013745 IKAA10013764 IKAA10013731
Rotational viscometers, ROTAVISC HELI complete series
Viskosimetre Rotationsviskosimetre
The ROTAVISC HELI series determines the viscosity of liquids in all areas of application ranging from the laboratory to quality control. The four devices measure in different viscosity ranges.
  • Large TFT display allows for an intuitive menu guidance
  • Digital spirit level supports the correct set-up of the start-up condition
  • Ramp function simplifies repetitive tasks
  • Very high measurement accuracy
  • Stepless speed adjustment
  • Timer and graph function
  • HELISTAND moves the mounted viscometer and its rotating body slowly and uniformly up and down in the sample