Nucleic acid extraction, MPure™ Viral/Pathogen nucleic acids extraction kit, MP Biomedicals
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
117022130
ICNA117022130EA 2600 DKK
ICNA117022130
Nucleic acid extraction, MPure™ Viral/Pathogen nucleic acids extraction kit, MP Biomedicals
Nukleinsyrereagenser Nucleic Acid Purification Kits and Reagents
MPure™ Viral/Pathogen Nucleic Acids Extraction Kit is used with the MPure-12™ Automated Nucleic Acid Purification System for purification of viral DNA/RNA and pathogen DNA from swab samples (cell-rich samples).