GeneClean® PCR clean-up, turbo for PCR kit, MP Biomedicals
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
111103600 111103200 111103400
ICNA111103600EA 4420 DKK
ICNA111103600 ICNA111103200 ICNA111103400
GeneClean® PCR clean-up, turbo for PCR kit, MP Biomedicals
Nukleinsyrereagenser Nucleic Acid Purification Kits and Reagents
The GeneClean® Turbo for PCR Kit removes primers, nucleotides, enzymes, mineral oil, salts, and other impurities from DNA samples. DNA fragments purified with this kit are ready for direct use in all applications, including sequencing, microarray analysis, ligation and transformation, restriction digestion and labeling.
  • Optimised for PCR purification of DNA fragments of sizes 100 bp to 10 kb
  • Able to purify larger fragments due to less manipulation of bound DNA
  • Suitable for either vacuum or centrifugation processing
  • Turbo filter design saves time
The kit contains Glassmilk® embedded filters in unique Turbo cartridges for maximal efficiency and convenience. Glassmilk® filters are able to preferentially bind nucleic acids in the presence of high salts and low pH. Since the binding process is specific for nucleic acids, the bound molecules can be separated and purified from impurities by simple washing steps. The DNA is then eluted from the matrix in low salt buffer or water, ready to use for downstream applications.
Leveringsinformation: Kit contains GeneClean® Turbo salt solution, GeneClean® Turbo wash solution, GeneClean® Turbo cartridges, GeneClean® Turbo catch tubes and GeneClean® Turbo elution solution.