RNA Purification kits, RNAID® kit, MP Biomedicals
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
111007200
ICNA111007200EA 2350 DKK
ICNA111007200
RNA Purification kits, RNAID® kit, MP Biomedicals
Nukleinsyrereagenser Nucleic Acid Purification Kits and Reagents
The RNaid® Kit is designed to isolate and purify RNA (75 bases to 10 kb) from native and denaturing agarose and polyacrylamide gels.
Ideal for removal of unincorporated nucleotides, buffer salts, and other reagents from enzyme reactions. The purified RNA is ready for reverse transcription and PCR, labeling and microarray analysis, and other common applications.