Dehydrated culture media, terrific broth, MP Biomedicals
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
091012017
ICNA091012017EA 682 DKK
ICNA091012017
Dehydrated culture media, terrific broth, MP Biomedicals
Mikrobiologiske medier Dehydreret Mikrobiologisk medie
This dehydrated medium is used for molecular biology applications.
  • Ideal for bacteria cell culture
  • To be stored at room temperature