Trichoderma viride cellulase Y-C, MP Biomedicals

Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

08320961
ICNA08320961EA 7350 DKK
ICNA08320961
Trichoderma viride cellulase Y-C, MP Biomedicals
Enzymer

Cellulase Y-C removes cell walls from plant tissues in shorter incubation period without loss of biological activity of the materials.


  • Readily soluble in water
  • Ideal for protoplast preparation


It is an effective enzyme for use with pectolyase Y-23 for plant cell wall removal. It has been used in a study to assess the differential regulation of the cellulase transcription factors XYR1, ACE2, and ACE. It has also been used in a study to investigate the liquefaction of hydrothermally pretreated wheat straw at high-solids content.

Bestil nu

Specifikationer

Enzym navn Cellulase Y-C
Environmentally Preferable
Kilde
Species Trichoderma viride
CAS-nr 9012-54-8
Form Light brown powder
Storage conditions 2...8 °C
Enzym aktivitet 16000 u/g
pH 4,0 - 5,0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR