Kaliumchlorid 99, powder_crystalline_white Molecular_Biology

Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Potassiumchloride

0219484494 0219484490 0219484491
ICNA0219484494EA 3550 DKK
ICNA0219484494 ICNA0219484490 ICNA0219484491
Kaliumchlorid 99, powder_crystalline_white Molecular_Biology
Potassium chloride

Potassium chloride is a salt commonly used in the preparation of phosphate buffered saline and for the extraction and solubilization of proteins.It is a good potassium source. Also used in the hypotonic KCl-propidium iodide protocol for univariate analysis of metaphase chromosomes; dissociation of eukaryotic ribosomes into subunits by high concentrations of potassium (0.3-1.0 M); preparation of mitochondria, provides an ionic osmotic support for those tissues that assume a gelatinous consistency upon homogenization.


  • Molecular Biology Reagent Grade
  • Presentation: White Crystalline Powder
  • Soluble in water (1 g/3 mL - clear, colorless solution), boiling water (1 g/1.8 mL), glycerol (1 g/14 mL), or ethanol (1 g/250 mL); insoluble in ether or acetone; hydrochloric acid, sodium or magnesium chlorides diminish its solubility in water.

Formel: KCl
Molekylvægt: 74.55 g/mol
Kogepunkt: ∼1500 °C (lit.)
Smeltepunkt: ∼770 °C (lit.)
Massefylde: 1.984 g/cm³ (lit.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011360
CAS-nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Bestil nu

Specifikationer Testresultater

Identity Test Passes
Purity ≥99%
pH (5% aq soln) 5.4-8.6
Insoluble Matter ≤0.005%
Iodide ≤0.002%
Bromide ≤0.01%
Chlorate & Nitrate ≤0.003%
Phosphate ≤5ppm
Iron ≤3ppm
Calcium ≤0.002%
Magnesium ≤0.001%
Sodium ≤0.005%
Loss on Drying (105°C/2h) ≤1.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR