Glycin

Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Aminoacetic acid, Glycocoll, H-Gly-OH

0219468101
ICNA0219468101EA 1110 DKK
ICNA0219468101
Glycin
Glycin
Formel: NH₂CH₂COOH
Kogepunkt: 181 °C (1013 hPa)
Smeltepunkt: 83 °C
Massefylde: 1,64 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008131
CAS-nummer: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR