Benzoesyre 99,5, powder ACS

Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

0219141290 0219141291 0219141280
ICNA0219141290EA 620 DKK
ICNA0219141290 ICNA0219141291 ICNA0219141280
Benzoesyre 99,5, powder ACS
Benzoesyre

Insoluble in methanol.


Insoluble in methanol: ≤0,005%
Chlorine compounds: ≤0,005%
Sulphur compounds: ≤0,002%

Formel: C₇H₆O₂
Molekylvægt: 122.12 g/mol
Kogepunkt: 249
Smeltepunkt: 121…123 °C
Massefylde: 1.316 g/cm³ (20 °C)
Flammepunkt: 121 °C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002398
CAS-nummer: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Index: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Form White Powder
Storage Store at room temperature (15 to 30 °C)
Heavy Metals ≤5 ppm
Residue on Ignition ≤0,005%
Freezing Point 122 to 123 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR