γ-L-Glutamyl-p-nitroanilide hydrochloride, beige powder

Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-(4-Nitrophenyl)-L-glutamine monohydrochloride, L-Glutamic acid -(p-nitroanilide) hydrochloride, L-2-Amino-4'-nitroglutaranilic acid hydrochloride

0215149501 02151495.2 0215149505
ICNA0215149501EA 1480 DKK
ICNA0215149501 ICNA02151495.2 ICNA0215149505
γ-L-Glutamyl-p-nitroanilide hydrochloride, beige powder
γ-L-Glutamyl-p-nitroanilide hydrochloride

γ-L-Glutamyl-p-Nitroanilide hydrochloride is suitable for use as a substrate for g-Glutamyl transpeptidase.


Soluble in water (5 mg/ml - clear, light yellow solution) or 0,1 N HCl (1% w/v - clear, yellowish solution); soluble in methanol (100 mg/ml with slight heat and constant stirring).


Advarsel: Research Use Only (RUO).

Formel: C₁₁H₁₄ClN₃O₅
MDL Number: MFCD00039054
CAS-nummer: 67953-08-6

Bestil nu

Specifikationer Testresultater

Identity Test Passes
Optical Rotation (c=1, 0.1 N HCl) +30° ± 5°
UV/Vis: λmax (water) 317 ± 5 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR