α-Linolensyre 45 - 65%, liquid

Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: ALA, (9Z,12Z,15Z)-Octadeca-9,12,15-trienoic acid, Linolenic acid, 9Z,12Z,15Z-Octadecatrienoic acid, cis,cis,cis-9,12,15-Octadecatrienoic acid

0210218890 0210218880 0210218891
ICNA0210218890EA 2790 DKK
ICNA0210218890 ICNA0210218880 ICNA0210218891
α-Linolensyre 45 - 65%, liquid
α-Linolensyre

Linolenic acid belongs to the class of omega-3 fatty acids.

Formel: C₁₈H₃₀O₂
MDL Number: MFCD00065720
CAS-nummer: 463-40-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR