Terry tool tubing cutters
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60182-510
HYPE60182-510EA 4110 DKK
HYPE60182-510
Terry tool tubing cutters
Knive
Tubing cutter cuts ¹/₈" stainless steel tubing.
Produce clean, 90° cuts of ¹/₈" stainless steel tubing.