Circulating cooling thermostats with natural refrigerant, Minichiller and Unichillers, with OLÉ controller
Leverandør: HUBER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3006.0091.98 3006.0089.98 3006.0090.98 3006.0092.98 3009.0090.98 3010.0050.98 3012.0120.98 3012.0126.98 3012.0142.98 3010.0103.98 3012.0260.98 3052.0012.98 3009.0244.98 3052.0018.98 3051.0020.98 3065.0001.98 3066.0002.98 3066.0003.98 3066.0004.98 3067.0001.98 3009.0200.98 3052.0020.98 3050.0003.98 3052.0024.98 3010.0130.98 3010.0124.98 3051.0018.98 3050.0012.98 3050.0026.98 3051.0003.98 3066.0001.98 3012.0263.98 3079.0006.98
HUBE3006.0091.98EA 43230 DKK
HUBE3006.0091.98 HUBE3006.0089.98 HUBE3006.0090.98 HUBE3006.0092.98 HUBE3009.0090.98 HUBE3010.0050.98 HUBE3012.0120.98 HUBE3012.0126.98 HUBE3012.0142.98 HUBE3010.0103.98 HUBE3012.0260.98 HUBE3052.0012.98 HUBE3009.0244.98 HUBE3052.0018.98 HUBE3051.0020.98 HUBE3065.0001.98 HUBE3066.0002.98 HUBE3066.0003.98 HUBE3066.0004.98 HUBE3067.0001.98 HUBE3009.0200.98 HUBE3052.0020.98 HUBE3050.0003.98 HUBE3052.0024.98 HUBE3010.0130.98 HUBE3010.0124.98 HUBE3051.0018.98 HUBE3050.0012.98 HUBE3050.0026.98 HUBE3051.0003.98 HUBE3066.0001.98 HUBE3012.0263.98 HUBE3079.0006.98
Circulating cooling thermostats with natural refrigerant, Minichiller and Unichillers, with OLÉ controller
Cirkulatorsystemer Lukkede cirkulatorsystemer Køletermostatbade
Many applications require a reliable source of cooling. Circulating chillers in the Unichiller® range offer an ideal solution for environmentally friendly and economical cooling in laboratory and industry. There are over 50 air and water cooled models to choose from, with cooling powers from 0,3 to 50 kW. Efficient energy management ensures low operating costs and reduced use of valuable fresh water. These circulating chillers are a resource saving solution which give a quick return on investment.
  • Robust stainless steel construction, ideal for simple applications
  • Large OLED display