Thermostatic baths for viscometers, CC®
Leverandør: HUBER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2001.0006.01 2048.0001.01
HUBE2001.0006.01EA 30500 DKK
HUBE2001.0006.01 HUBE2048.0001.01
Thermostatic baths for viscometers, CC®
Bade Viskosimeterbade
Visco thermostats are mainly used with capillary viscometers or densitometers for the measurement applications. These are offers the highest temperature stability for optimal temperature distribution in the thermostat bath.
  • Pilot ONE controller with plug and play technology
  • Precise temperature control
  • Overtemperature and low-level protection
  • Pt100 sensor connection
  • Powerful pressure/suction pump
  • Two-point calibration
The units offer the highest temperature stability for optimal temperature distribution in the thermostat bath. For operation below orMore Product Information