Pressure booster
Leverandør: HUBER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1085.0001.00
HUBE1085.0001.00EA 23290 DKK
HUBE1085.0001.00
Pressure booster
Pumper Kompressorer
The stainless steel Unipump® is designed to compensate for pressure loss in external systems.
  • Operation either manual (on/off) or external via 3-line-connector
The Unipump® is connected in series with the pump of the Unistat, Unichiller or Compatible Control circulator and can be controlled via the voltfree contact of the Com.Gate (Option).
Pump connection: M24×1,5
Max. pump capacities: 55 L/min, 1000 mbar
Bestillingsoplysninger: Accessories included. There are also some optional accessories.