Selenium AAS Standards, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E4067-100ML E4067-500ML E4118-100ML
HONCE4067-100MLEA 254 DKK
HONCE4067-100ML HONCE4067-500ML HONCE4118-100ML
Selenium AAS Standards, (CRMs), Fluka™
Standarder Kationiske standarder Selenium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificeringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).