Acetate Standard for Ion Chromatography, (CRM), Fluka™
Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E3000-100ML
HONCE3000-100MLEA 408 DKK
HONCE3000-100ML
Acetate Standard for Ion Chromatography, (CRM), Fluka™
Standarder Anioniske standarder Acetate Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Certificeringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).