Cadmium ICP & ICP-MS standard, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E1309-100ML
HONCE1309-100MLEA 722 DKK
HONCE1309-100ML
Cadmium ICP & ICP-MS standard, (CRMs), Fluka™
Standarder Kationiske standarder Cadmium Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Certificeringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).